Assay Details
| Gene Symbol: spe-45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322777.2 | NP_001309449.1 | 5 - 6 | 703 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spe-45| Gene Symbol: spe-45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322777.2 | NP_001309449.1 | 5 - 6 | 703 | 86 | |
Set Membership: |
|