Assay Details
| Gene Symbol: ipmk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070001.4 | NP_502402.3 | 4 - 5 | 358 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ipmk-1| Gene Symbol: ipmk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070001.4 | NP_502402.3 | 4 - 5 | 358 | 94 | |
Set Membership: |
|