Assay Details
Gene Symbol: efa-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070015.6 | NP_502416.1 | 4 - 5 | 592 | 109 | |
NM_070016.6 | NP_502417.1 | 4 - 5 | 592 | 109 | ||
GenBank mRNA | EF473217.1 | - | 3 - 4 | 535 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
