Assay Details
| Gene Symbol: wdfy-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268727.2 | NP_001255656.1 | 14 - 15 | 2810 | 100 | |
| NM_001268728.4 | NP_001255657.1 | 14 - 15 | 2791 | 100 | ||
| NM_001268729.3 | NP_001255658.1 | 14 - 15 | 2812 | 100 | ||
| NM_001268730.3 | NP_001255659.1 | 14 - 15 | 2724 | 100 | ||
More Information
Set Membership: |
|

wdfy-3
Genomic Map