Assay Details
Gene Symbol: wdfy-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268727.2 | NP_001255656.1 | 33 - 34 | 9403 | 101 | |
NM_001268728.4 | NP_001255657.1 | 33 - 34 | 9390 | 101 | ||
NM_001268729.3 | NP_001255658.1 | 33 - 34 | 9417 | 101 | ||
NM_001268730.3 | NP_001255659.1 | 33 - 34 | 9323 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
