Assay Details
| Gene Symbol: prmt-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047525.5 | NP_001040990.1 | 1 - 2 | 164 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prmt-9| Gene Symbol: prmt-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047525.5 | NP_001040990.1 | 1 - 2 | 164 | 66 | |
Set Membership: |
|