Assay Details
| Gene Symbol: inx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070034.5 | NP_502435.1 | 5 - 6 | 809 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

inx-6| Gene Symbol: inx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070034.5 | NP_502435.1 | 5 - 6 | 809 | 100 | |
Set Membership: |
|