Assay Details
| Gene Symbol: srf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268747.2 | NP_001255676.1 | 2 - 3 | 201 | 100 | |
| NM_001268748.3 | NP_001255677.1 | 1 - 2 | 65 | 100 | ||
More Information
Set Membership: |
|

srf-3
Genomic Map