Assay Details
| Gene Symbol: srf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268747.2 | NP_001255676.1 | 4 - 5 | 451 | 67 | |
| NM_001268748.3 | NP_001255677.1 | 3 - 4 | 315 | 67 | ||
| NM_001268749.3 | NP_001255678.1 | 1 - 2 | 63 | 67 | ||
More Information
Set Membership: |
|

srf-3
Genomic Map