Assay Details
| Gene Symbol: srf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268747.2 | NP_001255676.1 | 5 - 6 | 540 | 68 | |
| NM_001268748.3 | NP_001255677.1 | 4 - 5 | 404 | 68 | ||
| NM_001268749.3 | NP_001255678.1 | 2 - 3 | 152 | 68 | ||
More Information
Set Membership: |
|

srf-3
Genomic Map