Assay Details
| Gene Symbol: gcy-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070048.7 | NP_502449.2 | 2 - 3 | 421 | 124 | |
| GenBank mRNA | AB201389.1 | - | 2 - 3 | 381 | 124 | |
More Information
Set Membership: |
|

gcy-18
Genomic Map