Assay Details
| Gene Symbol: dod-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070073.7 | NP_502474.1 | 2 - 3 | 1037 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dod-24| Gene Symbol: dod-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070073.7 | NP_502474.1 | 2 - 3 | 1037 | 112 | |
Set Membership: |
|