Assay Details
| Gene Symbol: dod-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268760.2 | NP_001255689.1 | 5 - 6 | 797 | 69 | |
| NM_001268761.3 | NP_001255690.1 | 4 - 5 | 594 | 69 | ||
More Information
Set Membership: |
|

dod-22
Genomic Map