Assay Details
| Gene Symbol: spe-43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268773.1 | NP_001255702.1 | 2 - 3 | 162 | 60 | |
| NM_001268774.1 | NP_001255703.1 | 2 - 3 | 162 | 60 | ||
More Information
Set Membership: |
|

spe-43
Genomic Map