Assay Details
| Gene Symbol: msh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070130.8 | NP_502531.1 | 10 - 11 | 1297 | 79 | |
| GenBank mRNA | AF271389.1 | - | 10 - 11 | 1297 | 79 | |
More Information
Set Membership: |
|

msh-5
Genomic Map