Assay Details
| Gene Symbol: msh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380506.1 | NP_001368472.1 | 2 - 3 | 196 | 92 | |
| NM_070130.8 | NP_502531.1 | 12 - 13 | 1534 | 92 | ||
| GenBank mRNA | AF271389.1 | - | 12 - 13 | 1534 | 92 | |
More Information
Set Membership: |
|

msh-5
Genomic Map