Assay Details
| Gene Symbol: msh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380506.1 | NP_001368472.1 | 6 - 7 | 1649 | 90 | |
| NM_070130.8 | NP_502531.1 | 16 - 17 | 2987 | 90 | ||
| GenBank mRNA | AF271389.1 | - | 16 - 17 | 2987 | 90 | |
More Information
Set Membership: |
|

msh-5
Genomic Map