Assay Details
| Gene Symbol: msh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070130.8 | NP_502531.1 | 4 - 5 | 598 | 84 | |
| GenBank mRNA | AF271389.1 | - | 4 - 5 | 598 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

msh-5| Gene Symbol: msh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070130.8 | NP_502531.1 | 4 - 5 | 598 | 84 | |
| GenBank mRNA | AF271389.1 | - | 4 - 5 | 598 | 84 | |
Set Membership: |
|