Assay Details
| Gene Symbol: lin-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372945.2 | NP_001360500.1 | 1 - 2 | 154 | 63 | |
| NM_070143.4 | NP_502544.1 | 1 - 2 | 148 | 63 | ||
More Information
Set Membership: |
|

lin-54
Genomic Map