Assay Details
| Gene Symbol: lin-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372946.3 | NP_001360501.1 | 2 - 3 | 291 | 96 | |
| NM_070143.4 | NP_502544.1 | 2 - 3 | 294 | 96 | ||
More Information
Set Membership: |
|

lin-54
Genomic Map