Assay Details
| Gene Symbol: cpsf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070152.6 | NP_502553.2 | 9 - 10 | 2081 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpsf-3| Gene Symbol: cpsf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070152.6 | NP_502553.2 | 9 - 10 | 2081 | 86 | |
Set Membership: |
|