Assay Details
| Gene Symbol: rec-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268798.2 | NP_001255727.1 | 2 - 3 | 252 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rec-8| Gene Symbol: rec-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268798.2 | NP_001255727.1 | 2 - 3 | 252 | 121 | |
Set Membership: |
|