Assay Details
| Gene Symbol: rec-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268798.2 | NP_001255727.1 | 6 - 7 | 2157 | 66 | |
| NM_001268799.3 | NP_001255728.1 | 2 - 3 | 1575 | 66 | ||
More Information
Set Membership: |
|

rec-8
Genomic Map