Assay Details
| Gene Symbol: rla-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383231.1 | 3 - 4 | 512 | 80 | ||
| Gene Symbol: agt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047589.3 | NP_001041054.1 | 1 - 2 | 49 | 80 | |
More Information
Set Membership: |
|

rla-2
Genomic Map