Assay Details
| Gene Symbol: agt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047589.3 | NP_001041054.1 | 2 - 3 | 192 | 94 | |
| NM_001047590.4 | NP_001041055.1 | 1 - 2 | 66 | 94 | ||
| Gene Symbol: rla-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383231.1 | 4 - 5 | 655 | 94 | ||
More Information
Set Membership: |
|

agt-1
Genomic Map