Assay Details
Gene Symbol: rab-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383234.1 | 7 - 8 | 1078 | 65 |
Gene Symbol: Y62E10A.20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047588.5 | NP_001041053.1 | 7 - 8 | 1089 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
