Assay Details
| Gene Symbol: Y45F10B.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070218.1 | NP_502619.1 | 1 - 2 | 233 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y45F10B.12| Gene Symbol: Y45F10B.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070218.1 | NP_502619.1 | 1 - 2 | 233 | 145 | |
Set Membership: |
|