Assay Details
| Gene Symbol: tor-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070282.5 | NP_502683.2 | 2 - 3 | 526 | 171 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tor-2| Gene Symbol: tor-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070282.5 | NP_502683.2 | 2 - 3 | 526 | 171 | |
Set Membership: |
|