Assay Details
| Gene Symbol: irld-56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070286.3 | NP_502687.3 | 6 - 7 | 1292 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

irld-56| Gene Symbol: irld-56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070286.3 | NP_502687.3 | 6 - 7 | 1292 | 70 | |
Set Membership: |
|