Assay Details
| Gene Symbol: cfp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380528.1 | NP_001367055.1 | 3 - 4 | 845 | 67 | |
| NM_001380529.1 | NP_001367271.1 | 3 - 4 | 916 | 67 | ||
| NM_001380530.1 | NP_001367237.1 | 2 - 3 | 559 | 67 | ||
| NM_001380531.1 | NP_001367272.1 | 1 - 2 | 190 | 67 | ||
| NM_001383244.1 | 4 - 5 | 1129 | 67 | |||
More Information
Set Membership: |
|

cfp-1
Genomic Map