Assay Details
| Gene Symbol: R05A10.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070315.5 | NP_502716.2 | 2 - 3 | 248 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

R05A10.6| Gene Symbol: R05A10.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070315.5 | NP_502716.2 | 2 - 3 | 248 | 67 | |
Set Membership: |
|