Assay Details
| Gene Symbol: dmd-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383245.1 | 2 - 3 | 229 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383245.1 transcripts.
Set Membership: |
|

dmd-6
Genomic Map