Assay Details
| Gene Symbol: dmd-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268887.3 | NP_001255816.1 | 2 - 3 | 263 | 60 | |
| NM_001268888.3 | NP_001255817.1 | 1 - 2 | 206 | 60 | ||
| NM_001383245.1 | 6 - 7 | 896 | 60 | |||
More Information
Set Membership: |
|

dmd-6
Genomic Map