Assay Details
| Gene Symbol: mrps-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070381.3 | NP_502782.2 | 4 - 5 | 554 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrps-7| Gene Symbol: mrps-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070381.3 | NP_502782.2 | 4 - 5 | 554 | 62 | |
Set Membership: |
|