Assay Details
| Gene Symbol: eps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028350.6 | NP_001023521.2 | 14 - 15 | 2417 | 91 | |
| NM_001028351.5 | NP_001023522.2 | 11 - 12 | 1879 | 91 | ||
| NM_001047582.5 | NP_001041047.1 | 16 - 17 | 2709 | 91 | ||
| NM_001306998.2 | NP_001293927.1 | 17 - 18 | 2783 | 91 | ||
| NM_001306999.3 | NP_001293928.1 | 16 - 17 | 2543 | 91 | ||
| NM_001307000.3 | NP_001293929.1 | 15 - 16 | 2492 | 91 | ||
| NM_001307001.3 | NP_001293930.1 | 12 - 13 | 1952 | 91 | ||
| NM_001307002.3 | NP_001293931.1 | 4 - 5 | 419 | 91 | ||
| NM_001372963.2 | NP_001360610.1 | 17 | 2711 | 91 | ||
| NM_001372964.2 | NP_001360611.1 | 16 | 2636 | 91 | ||
| NM_070402.6 | NP_502803.3 | 15 - 16 | 2468 | 91 | ||
| GenBank mRNA | AY611484.1 | - | 16 - 17 | 2633 | 91 | |
More Information
Set Membership: |
|

eps-8
Genomic Map