Assay Details
| Gene Symbol: eps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028350.6 | NP_001023521.2 | 1 - 2 | 188 | 83 | |
| NM_001047582.5 | NP_001041047.1 | 3 - 4 | 480 | 83 | ||
| NM_001047583.7 | NP_001041048.1 | 3 - 4 | 486 | 83 | ||
| NM_001306998.2 | NP_001293927.1 | 3 - 4 | 479 | 83 | ||
| NM_001306999.3 | NP_001293928.1 | 2 - 3 | 239 | 83 | ||
| NM_001307000.3 | NP_001293929.1 | 1 - 2 | 188 | 83 | ||
| NM_001372963.2 | NP_001360610.1 | 3 | 407 | 83 | ||
| NM_001372964.2 | NP_001360611.1 | 3 | 407 | 83 | ||
| NM_070402.6 | NP_502803.3 | 2 - 3 | 239 | 83 | ||
| GenBank mRNA | AY611484.1 | - | 3 - 4 | 404 | 83 | |
| AY611485.1 | - | 3 - 4 | 404 | 83 | ||
More Information
Set Membership: |
|

eps-8
Genomic Map