Assay Details
| Gene Symbol: pkd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070437.5 | NP_502838.3 | 8 - 9 | 2083 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pkd-2| Gene Symbol: pkd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070437.5 | NP_502838.3 | 8 - 9 | 2083 | 98 | |
Set Membership: |
|