Assay Details
| Gene Symbol: gln-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047544.7 | NP_001041009.1 | 1 - 2 | 179 | 69 | |
| NM_001047545.4 | NP_001041010.1 | 1 - 2 | 170 | 69 | ||
More Information
Set Membership: |
|

gln-3
Genomic Map