Assay Details
| Gene Symbol: srz-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028097.3 | NP_001023268.2 | 4 - 5 | 745 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srz-104| Gene Symbol: srz-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028097.3 | NP_001023268.2 | 4 - 5 | 745 | 108 | |
Set Membership: |
|