Assay Details
| Gene Symbol: sta-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129343.3 | NP_001122815.1 | 5 - 6 | 1674 | 89 | |
| NM_070573.6 | NP_502974.2 | 5 - 6 | 1674 | 89 | ||
| GenBank mRNA | AY943817.1 | - | 5 - 6 | 1696 | 89 | |
More Information
Set Membership: |
|

sta-1
Genomic Map