Assay Details
| Gene Symbol: map-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268977.4 | NP_001255906.1 | 4 - 5 | 688 | 68 | |
| NM_001268978.4 | NP_001255907.1 | 5 - 6 | 718 | 68 | ||
More Information
Set Membership: |
|

map-2
Genomic Map