Assay Details
| Gene Symbol: map-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268977.4 | NP_001255906.1 | 5 - 6 | 1246 | 70 | |
| NM_001268978.4 | NP_001255907.1 | 6 - 7 | 1276 | 70 | ||
More Information
Set Membership: |
|

map-2
Genomic Map