Assay Details
| Gene Symbol: sfa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070632.6 | NP_503033.1 | 3 - 4 | 666 | 82 | |
| GenBank mRNA | AJ243905.1 | - | 3 - 4 | 666 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sfa-1| Gene Symbol: sfa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070632.6 | NP_503033.1 | 3 - 4 | 666 | 82 | |
| GenBank mRNA | AJ243905.1 | - | 3 - 4 | 666 | 82 | |
Set Membership: |
|