Assay Details
| Gene Symbol: sfa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070632.6 | NP_503033.1 | 5 - 6 | 891 | 113 | |
| NR_132466.1 | 4 - 5 | 818 | 113 | |||
| GenBank mRNA | AJ243905.1 | - | 5 - 6 | 891 | 113 | |
More Information
Set Membership: |
|

sfa-1
Genomic Map