Assay Details
| Gene Symbol: sfa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070632.6 | NP_503033.1 | 7 - 8 | 1763 | 97 | |
| NR_132466.1 | 6 - 7 | 1690 | 97 | |||
| GenBank mRNA | AJ243905.1 | - | 7 - 8 | 1763 | 97 | |
More Information
Set Membership: |
|

sfa-1
Genomic Map