Assay Details
| Gene Symbol: itsn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268998.3 | NP_001255927.1 | 1 - 2 | 151 | 75 | |
| NM_001313049.4 | NP_001299978.1 | 1 - 2 | 151 | 75 | ||
More Information
Set Membership: |
|

itsn-1
Genomic Map