Assay Details
| Gene Symbol: skr-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF440513.1 | - | 1 | 521 | 144 | |
| U60980.1 | - | 1 | 529 | 144 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

skr-12| Gene Symbol: skr-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF440513.1 | - | 1 | 521 | 144 | |
| U60980.1 | - | 1 | 529 | 144 | ||
Set Membership: |
|