Assay Details
| Gene Symbol: snf-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070676.4 | NP_503077.2 | 6 - 7 | 1158 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

snf-9| Gene Symbol: snf-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070676.4 | NP_503077.2 | 6 - 7 | 1158 | 97 | |
Set Membership: |
|