Assay Details
| Gene Symbol: dlat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269021.4 | NP_001255950.1 | 2 - 3 | 122 | 85 | |
| Gene Symbol: C30H6.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383265.1 | 1 - 2 | 44 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383265.1 transcripts.
Set Membership: |
|

dlat-2
Genomic Map