Assay Details
| Gene Symbol: dlat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269021.4 | NP_001255950.1 | 7 - 8 | 928 | 63 | |
| NR_068536.1 | 6 - 7 | 859 | 63 | |||
| Gene Symbol: C30H6.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383265.1 | 6 - 7 | 847 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383265.1 transcripts.
Set Membership: |
|

dlat-2
Genomic Map